Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bckdk
Entrez Gene ID
12041
Gene Name
branched chain ketoacid dehydrogenase kinase
Gene Aliases
AI327402
Chromosome Location
Chr.7: 127904073 - 127909664 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009739.3 NP_033869.1 6 - 7 808 79
GenBank mRNA AF043070.1 - 5 - 6 649 79
AK075602.1 - 6 - 7 734 79
AK145479.1 - 6 - 7 779 79
AK150607.1 - 6 - 7 800 79
AK150723.1 - 6 - 7 800 79
AK167519.1 - 6 - 7 800 79
BC004077.1 - 3 - 4 236 79
BC046595.1 - 6 - 7 778 79

View Full Product Details