Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajc9
Entrez Gene ID
108671
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 9
Gene Aliases
5330419I01Rik, AU020082
Chromosome Location
Chr.14: 20384638 - 20388910 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134081.5 NP_598842.1 2 - 3 453 87
GenBank mRNA AK043725.1 - 2 - 3 423 87
AK075979.1 - 2 - 3 414 87
AK077324.1 - 2 - 3 423 87
AK153033.1 - 2 - 3 413 87
AK162075.1 - 2 - 3 328 87
BC014686.1 - 2 - 3 415 87
BC023787.1 - 2 - 3 411 87
BC027012.1 - 2 - 3 406 87
CX238858.1 - 2 - 3 453 87

View Full Product Details