Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp1r12b
Entrez Gene ID
329251
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 12B
Gene Aliases
1810037O03Rik, 9530009M10Rik, AI132431, Mypt2
Chromosome Location
Chr.1: 134754665 - 134955965 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081307.1 NP_001074776.1 8 - 9 1296 94
GenBank mRNA AK046012.1 - 8 - 9 1285 94
AK046167.1 - 8 - 9 1285 94
AK080855.1 - 8 - 9 1285 94
AK141375.1 - 8 - 9 1285 94
BC145746.1 - 8 - 9 1207 94

View Full Product Details