Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc7a11
Entrez Gene ID
26570
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 11
Gene Aliases
9930009M05Rik, AI451155, sut, xCT
Chromosome Location
Chr.3: 50364936 - 50499087 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011990.2 NP_036120.1 5 - 6 1099 125
GenBank mRNA AB022345.1 - 5 - 6 1066 125
AK076322.1 - 5 - 6 1074 125
AK089202.1 - 5 - 6 1072 125
AK162233.1 - 5 - 6 874 125
AK163168.1 - 5 - 6 1073 125
AK165599.1 - 5 - 6 1075 125
AK171192.1 - 5 - 6 1097 125
AK171252.1 - 5 - 6 1073 125
AY766236.1 - 5 - 6 1066 125
AY766237.1 - - 1066 125
BC141402.1 - 5 - 6 996 125
BC141408.1 - 5 - 6 996 125

View Full Product Details