Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gm2a
Entrez Gene ID
14667
Gene Name
GM2 ganglioside activator protein
Gene Aliases
AA408702, AW215435, GM2-AP, SAP-3
Chromosome Location
Chr.11: 55097985 - 55113029 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010299.3 NP_034429.1 2 - 3 423 64
GenBank mRNA AK149642.1 - 2 - 3 315 64
AK152256.1 - 2 - 3 315 64
BC004651.1 - 2 - 3 291 64
CT010289.1 - 2 - 3 244 64
L19526.1 - 2 - 3 257 64
U09816.1 - 2 - 3 300 64

View Full Product Details