The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172644.3 | NP_766232.1 | 3 - 4 | 812 | 68 |
GenBank mRNA | AK030952.1 | - | 3 - 4 | 801 | 68 |
AK033634.1 | - | 2 - 3 | 1635 | 68 | |
AK037337.1 | - | 3 - 4 | 831 | 68 | |
AK048454.1 | - | 3 - 4 | 789 | 68 | |
AK138809.1 | - | 3 - 4 | 813 | 68 | |
AK154133.1 | - | 3 - 4 | 841 | 68 | |
AK154348.1 | - | 3 - 4 | 841 | 68 | |
BC057943.1 | - | 3 - 4 | 836 | 68 | |
BC076606.1 | - | 3 - 4 | 804 | 68 |