Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oxt
Entrez Gene ID
18429
Gene Name
oxytocin
Gene Aliases
OT, Oxy
Chromosome Location
Chr.2: 130574967 - 130577054 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011025.4 NP_035155.1 1 - 2 163 67
GenBank mRNA AF483508.1 - 1 - 2 122 67
AF483509.1 - 1 - 2 122 67
AK003042.1 - 1 - 2 160 67
AW455577.1 - 2 - 3 67
BC117030.1 - 1 - 2 142 67
BC117032.1 - 1 - 2 142 67

View Full Product Details