Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eci2
Entrez Gene ID
23986
Gene Name
enoyl-Coenzyme A delta isomerase 2
Gene Aliases
Acbd2, Drs1, Hca88, Peci
Chromosome Location
Chr.13: 34977748 - 35027089 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110331.1 NP_001103801.1 6 - 7 747 82
NM_011868.3 NP_035998.2 6 - 7 931 82
NR_073427.1 6 - 7 737 82
GenBank mRNA AF153613.1 - 6 - 7 903 82
AK009478.1 - 6 - 7 731 82
AK170778.1 - 6 - 7 716 82
BC001983.1 - 6 - 7 641 82

View Full Product Details