The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029232.2 | NP_083508.1 | 10 - 11 | 810 | 111 |
GenBank mRNA | AK016214.1 | - | 10 - 11 | 801 | 111 |
BC050803.1 | - | 10 - 11 | 810 | 111 |