Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Armcx6
Entrez Gene ID
278097
Gene Name
armadillo repeat containing, X-linked 6
Gene Aliases
AW060994
Chromosome Location
Chr.X: 134748447 - 134756342 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007578.2 NP_001007579.1 1 - 2 93 75
GenBank mRNA AK148507.1 - 1 - 2 92 75
AK160766.1 - 1 - 2 75
BC027264.1 - 1 - 2 86 75
BY777182.1 - 1 - 2 73 75

View Full Product Details