Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arvcf
Entrez Gene ID
11877
Gene Name
armadillo repeat gene deleted in velo-cardio-facial syndrome
Gene Aliases
-
Chromosome Location
Chr.16: 18348182 - 18407076 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272028.1 NP_001258957.1 4 - 4 1097 79
NM_001272029.1 NP_001258958.1 5 - 5 1248 79
NM_001272030.1 NP_001258959.1 4 - 4 1097 79
NM_001272031.1 NP_001258960.1 3 - 3 840 79
NM_001272032.1 NP_001258961.1 3 - 3 840 79
NM_033474.3 NP_258435.2 5 - 5 1248 79
GenBank mRNA AF286212.1 - 4 - 4 1097 79
AF286213.1 - 4 - 4 1097 79
AF286214.1 - 3 - 3 749 79
AF286215.1 - 3 - 3 749 79
AJ243418.1 - 4 - 4 882 79
AK044982.1 - 3 - 3 788 79
AK051627.1 - 3 - 3 796 79
AK053865.1 - 3 - 3 821 79
AK084886.1 - 5 - 5 1236 79
BC052393.1 - 5 - 5 1167 79
BC056980.1 - 5 - 5 1156 79

View Full Product Details