The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026871.1 | NP_081147.1 | 3 - 4 | 333 | 81 |
GenBank mRNA | AF356874.1 | - | 3 - 4 | 338 | 81 |
AK004497.1 | - | 3 - 4 | 335 | 81 | |
AK170135.1 | - | 2 - 3 | 1649 | 81 | |
BC086940.1 | - | 3 - 4 | 333 | 81 |