The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172755.3 | NP_766343.3 | 10 - 11 | 3643 | 70 |
GenBank mRNA | AF518875.1 | - | 10 - 11 | 3636 | 70 |