The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027182.2 | NP_081458.1 | 11 - 12 | 1320 | 71 |
GenBank mRNA | AK010336.1 | - | 11 - 12 | 1282 | 71 |
AK142463.1 | - | - | 1293 | 71 | |
AK146877.1 | - | 11 - 12 | 1321 | 71 | |
AK151568.1 | - | 11 - 12 | 1256 | 71 | |
BC023834.1 | - | 5 - 6 | 528 | 71 | |
BC064315.1 | - | 11 - 12 | 1243 | 71 | |
BC126946.1 | - | 11 - 12 | 1219 | 71 |