The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027182.2 | NP_081458.1 | 12 - 13 | 1387 | 74 |
GenBank mRNA | AK010336.1 | - | 12 - 13 | 1349 | 74 |
AK146877.1 | - | 12 - 13 | 1388 | 74 | |
AK151568.1 | - | 12 - 13 | 1323 | 74 | |
BC023834.1 | - | 6 - 7 | 595 | 74 | |
BC064315.1 | - | 12 - 13 | 1310 | 74 | |
BC126946.1 | - | 12 - 13 | 1286 | 74 |