Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trip13
Entrez Gene ID
69716
Gene Name
thyroid hormone receptor interactor 13
Gene Aliases
2410002G23Rik, D13Ertd328e
Chromosome Location
Chr.13: 73912462 - 73937767 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027182.2 NP_081458.1 7 - 8 864 113
GenBank mRNA AK010336.1 - 7 - 8 826 113
AK142463.1 - - 837 113
AK146877.1 - 7 - 8 865 113
AK151568.1 - 7 - 8 800 113
BC064315.1 - 7 - 8 787 113
BC126946.1 - 7 - 8 763 113

View Full Product Details