Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp4f16
Entrez Gene ID
70101
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 16
Gene Aliases
2310021J05Rik, AI746515
Chromosome Location
Chr.17: 32536545 - 32551798 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK083886.1 - 2 - 3 283 106

View Full Product Details