Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nutm2
Entrez Gene ID
328250
Gene Name
NUT family member 2
Gene Aliases
Fam22, Gm806
Chromosome Location
Chr.13: 50467307 - 50475355 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033400.2 NP_001028572.1 4 - 5 1117 80
GenBank mRNA AK133253.1 - 4 - 5 1118 80

View Full Product Details