Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Slco4c1
- Entrez Gene ID
-
227394
- Gene Name
- solute carrier organic anion transporter family, member 4C1
- Gene Aliases
-
9530051F04, C330017E21Rik, OATP-H, OATP-M1, OATP4C1, OATPX, PRO2176, SLC21A20, oatp-R
- Chromosome Location
-
Chr.1: 96818784 - 96872171 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK035459.1 |
- |
- |
1653 |
90 |
View Full Product Details