Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cxcl13
Entrez Gene ID
55985
Gene Name
chemokine (C-X-C motif) ligand 13
Gene Aliases
4631412M08Rik, ANGIE2, Angie, BCA-1, BLC, BLR1L, Scyb13
Chromosome Location
Chr.5: 95956939 - 95961068 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018866.2 NP_061354.1 4 - 4 753 91
GenBank mRNA AF044196.1 - 4 - 4 753 91
AK156025.1 - 4 - 4 767 91
AK160241.1 - 4 - 4 769 91
BC012965.1 - 4 - 4 752 91

View Full Product Details