Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mirlet7f-1
- Entrez Gene ID
-
387252
- Gene Name
- microRNA let7f-1
- Gene Aliases
-
Mirlet7f1, Mirnlet7f1, let-7f-1
- Chromosome Location
-
Chr.13: 48537829 - 48537917 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029731.1 |
|
1 - 1 |
47 |
88 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CB237456.1 |
- |
1 |
391 |
|
Chr. 13 - 48537261 - 48538259 [-] on Build GRCm38 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA let7d |
Mirlet7d |
Mirnlet7d, let-7d, mmu-let-7d |
-1.7kb
|
Sense |
Chr. - 48536012 - 48536114 on Build GRCm38 |
Downstream |
microRNA let7a-1 |
Mirlet7a-1 |
Let-7a, Mirnlet7a, Mirnlet7a-1, let-7a-1 |
262.0
|
Sense |
Chr. - 48538179 - 48538272 on Build GRCm38 |
View Full Product Details