Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNG
Entrez Gene ID
101115316
Gene Name
uracil DNA glycosylase
Gene Aliases
-
Chromosome Location
Chr.17: 73197798 - 73208557 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027956650.1 XP_027812451.1 6 - 7 838 90

View Full Product Details