Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALOX12B
Entrez Gene ID
101105929
Gene Name
arachidonate 12-lipoxygenase, 12R type
Gene Aliases
-
Chromosome Location
Chr.11: 35696055 - 35707346 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012678.4 XP_004012727.2 3 - 4 657 57

View Full Product Details