Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIGR
Entrez Gene ID
100144764
Gene Name
polymeric immunoglobulin receptor
Gene Aliases
-
Chromosome Location
Chr.12: 4329562 - 4347557 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013573.3 XP_004013622.2 7 - 8 2038 49

View Full Product Details