Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDCD2
Entrez Gene ID
101118111
Gene Name
programmed cell death 2
Gene Aliases
-
Chromosome Location
Chr.8: 98750290 - 98759327 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972699.1 XP_027828500.1 2 - 3 556 76

View Full Product Details