Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB8B
Entrez Gene ID
101113392
Gene Name
zinc finger and BTB domain containing 8B
Gene Aliases
-
Chromosome Location
Chr.2: 250755324 - 250771586 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965344.1 XP_027821145.1 2 - 3 1467 71

View Full Product Details