Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM71E2
Entrez Gene ID
101120774
Gene Name
family with sequence similarity 71 member E2
Gene Aliases
-
Chromosome Location
Chr.14: 66808143 - 66811961 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977985.1 XP_027833786.1 7 - 8 805 63

View Full Product Details