Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RABL3
Entrez Gene ID
101108873
Gene Name
RAB, member of RAS oncogene family like 3
Gene Aliases
-
Chromosome Location
Chr.1: 203397267 - 203435073 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002956.4 XP_004003005.1 1 - 2 94 70

View Full Product Details