Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD1
Entrez Gene ID
101114505
Gene Name
abhydrolase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 35737076 - 35744351 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005753.4 XP_004005802.1 8 - 9 1128 57

View Full Product Details