Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSTD2
Entrez Gene ID
101104459
Gene Name
thiosulfate sulfurtransferase like domain containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 52694031 - 52722300 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004234.4 XP_004004283.1 9 - 10 1411 66
XM_012120651.3 XP_011976041.1 9 - 10 1448 66

View Full Product Details