Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBLN7
Entrez Gene ID
101118347
Gene Name
fibulin 7
Gene Aliases
-
Chromosome Location
Chr.3: 112437579 - 112495277 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006187.4 XP_004006236.2 6 - 7 1097 112
XM_012173910.2 XP_012029300.2 5 - 6 738 112

View Full Product Details