Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NHEJ1
Entrez Gene ID
101113915
Gene Name
non-homologous end joining factor 1
Gene Aliases
-
Chromosome Location
Chr.2: 235838072 - 235928446 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012146435.2 XP_012001825.2 3 - 4 582 78

View Full Product Details