Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNDC5
Entrez Gene ID
101104971
Gene Name
fibronectin type III domain containing 5
Gene Aliases
-
Chromosome Location
Chr.2: 250418188 - 250426205 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093613.2 XP_014949099.2 4 - 5 480 63
XM_015093614.2 XP_014949100.2 4 - 5 480 63
XM_015093615.2 XP_014949101.2 4 - 5 480 63

View Full Product Details