Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR3G
Entrez Gene ID
101106586
Gene Name
RNA polymerase III subunit G
Gene Aliases
-
Chromosome Location
Chr.5: 95009860 - 95063950 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096190.2 XP_014951676.2 1 - 2 2595 59

View Full Product Details