Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LSMEM1
Entrez Gene ID
101112307
Gene Name
leucine rich single-pass membrane protein 1
Gene Aliases
C4H7orf53
Chromosome Location
Chr.4: 62702069 - 62712644 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007880.3 XP_004007929.1 3 - 4 875 75
XM_012176781.2 XP_012032171.1 4 - 5 896 75
XM_012176782.2 XP_012032172.1 4 - 5 893 75
XM_027968610.1 XP_027824411.1 4 - 5 990 75

View Full Product Details