Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHPF
Entrez Gene ID
101114088
Gene Name
chondroitin polymerizing factor
Gene Aliases
-
Chromosome Location
Chr.2: 236273892 - 236279305 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965230.1 XP_027821031.1 2 - 3 1167 83

View Full Product Details