Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFAP2
Entrez Gene ID
101110395
Gene Name
microfibril associated protein 2
Gene Aliases
-
Chromosome Location
Chr.2: 265095840 - 265102231 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965678.1 XP_027821479.1 5 - 6 1545 71
XM_027965679.1 XP_027821480.1 5 - 6 349 71
XM_027965680.1 XP_027821481.1 5 - 6 332 71
XM_027965681.1 XP_027821482.1 5 - 6 330 71

View Full Product Details