Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAS1L
Entrez Gene ID
101112288
Gene Name
LAS1 like ribosome biogenesis factor
Gene Aliases
-
Chromosome Location
Chr.X: 49650609 - 49670346 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022035.4 XP_004022084.1 5 - 6 957 89
XM_004022036.4 XP_004022085.1 5 - 6 939 89
XM_012106584.3 XP_011961974.1 5 - 6 940 89

View Full Product Details