Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TF
Entrez Gene ID
100009267
Gene Name
transferrin
Gene Aliases
TRF
Chromosome Location
Chr.14: 27729544 - 27758319 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101694.1 NP_001095164.1 9 - 10 1226 104
GenBank mRNA X58533.1 - - 1226 104

View Full Product Details