Gene Transcripts

Gene Symbol
Trim34
Entrez Gene ID
101241893
Gene Name
tripartite motif-containing 34
Gene Aliases
Chromosome Location
Chr. 1: 169433539 - 169445041 on Build Rnor_6.0
UniGene ID
Rn.212480
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276491.1 NP_001263420.1 7, 8 1236
GenBank mRNA CO563709.1 - 3, 4 431

View Full Product Details