Gene Transcripts

Gene Symbol
Trpm2
Entrez Gene ID
294329
Gene Name
transient receptor potential cation channel, subfamily M, member 2
Gene Aliases
Trpm2-predicted
Chromosome Location
Chr. 20: 11434043 - 11482880 on Build Rnor_6.0
UniGene ID
Rn.214495
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011559.1 NP_001011559.1 19, 20 2951
XM_006256248.2 XP_006256310.1 20, 21 3141
XM_006256249.2 XP_006256311.1 20, 21 3141
XM_008772811.1 XP_008771033.1 20, 21 3141
GenBank mRNA AB193179.1 - 19, 20 2982
AY749166.1 - 19, 20 2951

View Full Product Details