Gene Transcripts

Gene Symbol
Arhgap44
Entrez Gene ID
303222
Gene Name
Rho GTPase activating protein 44
Gene Aliases
RGD1305664
Chromosome Location
Chr. 10: 51501547 - 51669548 on Build Rnor_6.0
UniGene ID
Rn.17471
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246834.2 XP_006246896.1 5, 6 729
XM_006246835.2 XP_006246897.1 5, 6 729
XM_008767895.1 XP_008766117.1 5, 6 729
XM_008767896.1 XP_008766118.1 5, 6 729

View Full Product Details