Gene Transcripts

Gene Symbol
Asna1
Entrez Gene ID
288919
Gene Name
arsA arsenite transporter, ATP-binding, homolog 1 (bacterial)
Gene Aliases
Chromosome Location
Chr. 19: 26133024 - 26142641 on Build Rnor_6.0
UniGene ID
Rn.7354
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772367.1 XP_008770589.1 1 306
XM_008772368.1 XP_008770590.1 2 310
XM_008772369.1 XP_008770591.1 2 305

View Full Product Details