Gene Transcripts

Gene Symbol
Irs2
Entrez Gene ID
29376
Gene Name
insulin receptor substrate 2
Gene Aliases
4PS, IRS-2
Chromosome Location
Chr. 16: 83824515 - 83848569 on Build Rnor_6.0
UniGene ID
Rn.10718
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168633.1 NP_001162104.1 1 2410
GenBank mRNA AF050159.1 - 1 937

View Full Product Details