Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PWP1
Entrez Gene ID
705262
Gene Name
PWP1 homolog, endonuclein
Gene Aliases
-
Chromosome Location
Chr.11: 107370305 - 107395089 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015152683.2 XP_015008169.1 - 482 85
GenBank mRNA JU332714.1 - - 406 85
JU474143.1 - - 405 85

View Full Product Details