Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOPC
Entrez Gene ID
717841
Gene Name
golgi associated PDZ and coiled-coil motif containing
Gene Aliases
-
Chromosome Location
Chr.4: 21081675 - 21124820 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015137529.2 XP_014993015.1 - 909 84
XM_015137530.2 XP_014993016.1 - 884 84
GenBank mRNA JU322047.1 - - 841 84
JU322061.1 - - 865 84
JU472307.1 - - 848 84
JU472308.1 - - 824 84
JV066301.1 - - 844 84
JV637808.1 - - 864 84

View Full Product Details