Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HINT2
Entrez Gene ID
695521
Gene Name
histidine triad nucleotide binding protein 2
Gene Aliases
EGK_07559
Chromosome Location
Chr.15: 48391143 - 48393667 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266678.1 NP_001253607.1 - 396 84
XM_015117334.1 XP_014972820.1 - 830 84
GenBank mRNA JU323101.1 - - 396 84
JV725349.1 - - 401 84

View Full Product Details