Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBLN7
Entrez Gene ID
698702
Gene Name
fibulin 7
Gene Aliases
-
Chromosome Location
Chr.13: 15363314 - 15411850 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001087308.4 XP_001087308.2 - 1090 126
GenBank mRNA JV605969.1 - - 270 126

View Full Product Details