Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOWAHD
Entrez Gene ID
694278
Gene Name
sosondowah ankyrin repeat domain family member D
Gene Aliases
ANKRD58
Chromosome Location
Chr.X: 116045180 - 116046788 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193702.1 NP_001180631.1 - 1251 72

View Full Product Details