Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMX2
Entrez Gene ID
707928
Gene Name
H6 family homeobox 2
Gene Aliases
EGK_20148
Chromosome Location
Chr.9: 123579772 - 123588059 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194301.2 NP_001181230.1 - 519 100
XM_002805844.3 XP_002805890.1 - 334 100

View Full Product Details